Anti-ARHGAP12 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB1136
10690-020EA 427.45 USD
10690-020
Anti-ARHGAP12 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ARHGAP12.

Recommended Dilutions: ELISA: 1:100-1:1000

Type: Primary
Antigen: ARHGAP12
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR