Anti-CFLAR Mouse Monoclonal Antibody [clone: 5D8]

Supplier: Abnova
MAB2065
10707-700EA 329 USD
10707-700
Anti-CFLAR Mouse Monoclonal Antibody [clone: 5D8]
Antibodies
Mouse monoclonal antibody raised against partial recombinant CFLAR.

Type: Primary
Antigen: CFLAR
Clonality: Monoclonal
Clone: 5D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR