Anti-SMT3 Mouse Monoclonal Antibody [clone: 4F2.F5.G2]

Supplier: Abnova
MAB2093
10738-754EA 746.62 USD
10738-754
Anti-SMT3 Mouse Monoclonal Antibody [clone: 4F2.F5.G2]
Antibodies
Mouse monoclonal antibody raised against full length recombinant SMT3.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:2000; Immunohistochemistry: 1:1000

Type: Primary
Antigen: SMT3
Clonality: Monoclonal
Clone: 4F2.F5.G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR