Anti-YWHAQ Mouse Monoclonal Antibody [clone: AT1A1]

Supplier: Abnova
MAB2098
10744-980EA 329 USD
10744-980
Anti-YWHAQ Mouse Monoclonal Antibody [clone: AT1A1]
Antibodies
Mouse monoclonal antibody raised against partial recombinant YWHAQ.

Type: Primary
Antigen: YWHAQ
Clonality: Monoclonal
Clone: AT1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR