Anti-SCARB1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12123
10732-336EA 530.32 USD
10732-336
Anti-SCARB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Scarb1/Scarb2.

Recommended Dilutions: Immunocytochemistry: 1:50-1:1000; Immunofluorescence: 1:50-1:1000; Western Blot: 1:1000

Type: Primary
Antigen: SCARB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR