Anti-MED28 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22921
10725-274EA 428.55 USD
10725-274
Anti-MED28 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant MED28.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: MED28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR