Anti-LRRK2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12132
10724-080EA 530.32 USD
10724-080
Anti-LRRK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Lrrk2.

  • Recommended Dilutions: Western Blot: 2 μg/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2.5 μg/ml

Type: Primary
Antigen: LRRK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR