Anti-RBM27 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22930
10731-266EA 428.55 USD
10731-266
Anti-RBM27 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant RBM27.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: RBM27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR