Anti-AGGF1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12151
10688-432EA 653.85 USD
10688-432
Anti-AGGF1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Aggf1.

Recommended Dilutions: Immunohistochemistry: 1:250; Western Blot: 1:500

Type: Primary
Antigen: AGGF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR