Anti-FAM162B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22947
10713-668EA 428.55 USD
10713-668
Anti-FAM162B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant FAM162B.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: FAM162B
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR