Anti-IL1B Mouse Monoclonal Antibody [clone: 5C10]

Supplier: Abnova
MAB3006
10676-612EA 406.15 USD
10676-612
Anti-IL1B Mouse Monoclonal Antibody [clone: 5C10]
Antibodies
Mouse monoclonal antibody raised against recombinant IL1B.

Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: 5C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR