Anti-SHCBP1L Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22949
10703-900EA 428.55 USD
10703-900
Anti-SHCBP1L Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C1orf14.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: SHCBP1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR