Anti-PRR27 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22955
10704-350EA 527.69 USD
10704-350
Anti-PRR27 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C4orf40.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: PRR27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR