Anti-Procalcitonin Mouse Monoclonal Antibody [clone: 14C12]

Supplier: Abnova
MAB3015
10677-086EA 653.85 USD
10677-086
Anti-Procalcitonin Mouse Monoclonal Antibody [clone: 14C12]
Antibodies
Mouse monoclonal antibody raised against partial Procalcitonin.

Type: Primary
Antigen: Procalcitonin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 14C12
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR