Anti-Procalcitonin Mouse Monoclonal Antibody [clone: 22A11]

Supplier: Abnova
MAB3016
10677-088EA 653.85 USD
10677-088
Anti-Procalcitonin Mouse Monoclonal Antibody [clone: 22A11]
Antibodies
Mouse monoclonal antibody raised against partial Procalcitonin.

Type: Primary
Antigen: Procalcitonin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 22A11
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR