Anti-CAV1 Mouse Monoclonal Antibody [clone: 4D6]

Supplier: Abnova
MAB3768
10626-818EA 1960 USD
10626-818
Anti-CAV1 Mouse Monoclonal Antibody [clone: 4D6]
Antibodies
Mouse monoclonal antibody raised against partial recombinant CAV1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:200; Western Blot: 1:4000

Type: Primary
Antigen: CAV1
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR