Anti-BGLAP Mouse Monoclonal Antibody [clone: G12]

Supplier: Abnova
MAB3776
10691-172EA 527.69 USD
10691-172
Anti-BGLAP Mouse Monoclonal Antibody [clone: G12]
Antibodies
Mouse monoclonal antibody raised against native BGLAP.

Type: Primary
Antigen: BGLAP
Clonality: Monoclonal
Clone: G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR