Anti-ABCG2 Rat Monoclonal Antibody [clone: BXP-9]

Supplier: Abnova
MAB3780
10626-450EA 1060.7 USD
10626-450
Anti-ABCG2 Rat Monoclonal Antibody [clone: BXP-9]
Antibodies
Rat monoclonal antibody raised against partial recombinant Abcg2.

Recommended Dilutions: Immunocytochemistry: 1:20-1:50; Immunohistochemistry (Frozen sections): 1:20; Western Blot: 1:20-1:50

Type: Primary
Antigen: ABCG2
Clonality: Monoclonal
Clone: BXP-9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR