Anti-ARRDC4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23904
10692-586EA 428.55 USD
10692-586
Anti-ARRDC4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ARRDC4.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: ARRDC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR