Anti-PLAT Mouse Monoclonal Antibody [clone: H6C5]

Supplier: Abnova
MAB3791
10628-474EA 1277 USD
10628-474
Anti-PLAT Mouse Monoclonal Antibody [clone: H6C5]
Antibodies
Mouse monoclonal antibody raised against recombinant Plat.

Type: Primary
Antigen: PLAT
Clonality: Monoclonal
Clone: H6C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR