Anti-PLAT Mouse Monoclonal Antibody [clone: 629-2A153]

Supplier: Abnova
MAB3792
10628-470EA 1178.46 USD
10628-470
Anti-PLAT Mouse Monoclonal Antibody [clone: 629-2A153]
Antibodies
Mouse monoclonal antibody raised against recombinant PLAT.

Type: Primary
Antigen: PLAT
Clonality: Monoclonal
Clone: 629-2A153
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR