Anti-PLAU Mouse Monoclonal Antibody [clone: H77A10]

Supplier: Abnova
MAB3793
10628-480EA 1572.31 USD
10628-480
Anti-PLAU Mouse Monoclonal Antibody [clone: H77A10]
Antibodies
Mouse monoclonal antibody raised against recombinant Plau.

Type: Primary
Antigen: PLAU
Clonality: Monoclonal
Clone: H77A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR