Anti-COX1 Mouse Monoclonal Antibody

Supplier: Abnova
MAB3795
10730-560EA 527.69 USD
10730-560
Anti-COX1 Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against PTGS1.

Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry: 1:1000

Type: Primary
Antigen: COX1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR