Anti-SERPINE1 Mouse Monoclonal Antibody

Supplier: Abnova
MAB3802
10628-722EA 2827.69 USD
10628-722
Anti-SERPINE1 Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against SERPINE1 (inhibitory).

Type: Primary
Antigen: SERPINE1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR