Anti-SERPINE1 Mouse Monoclonal Antibody [clone: MA32K3]

Supplier: Abnova
MAB3804
10628-754EA 2226.15 USD
10628-754
Anti-SERPINE1 Mouse Monoclonal Antibody [clone: MA32K3]
Antibodies
Mouse monoclonal antibody raised against recombinant Serpine1.

Type: Primary
Antigen: SERPINE1
Clonality: Monoclonal
Clone: MA32K3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR