Anti-CENPE Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23920
10707-550EA 428.55 USD
10707-550
Anti-CENPE Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CENPE.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: CENPE
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR