Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYB5D2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23921
10709-686
Anti-CYB5D2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against recombinant CYB5D2.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: CYB5D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty