Anti-Antiplasmin (inhibitory) Mouse Monoclonal Antibody [clone: MAP4H9]

Supplier: Abnova
MAB3811
10626-574EA 1833.85 USD
10626-574
Anti-Antiplasmin (inhibitory) Mouse Monoclonal Antibody [clone: MAP4H9]
Antibodies
Mouse monoclonal antibody raised against Antiplasmin (inhibitory).

Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone: MAP4H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR