Anti-ATLF 0 Polyclonal Antibody

Supplier: Abnova
PAB13126
10689-262EA 428 USD
10689-262
Anti-ATLF 0 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Anthrax Lethal Factor.

Recommended Dilutions: ELISA: 1 ?g/ml

Type: Primary
Antigen: ATLF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: 0
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR