Anti-CD4 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: MEM-241]

Supplier: Abnova
MAB4514
10708-206EA 329 USD
10708-206
Anti-CD4 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: MEM-241]
Antibodies
Mouse monoclonal antibody raised against partial recombinant CD4.

Recommended Dilutions: Flow Cytometry: 10 ?g/ml

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: MEM-241
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR