Anti-HA Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13130
10690-572EA 428 USD
10690-572
Anti-HA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Avian Influenza Hemagglutinin.

Recommended Dilutions: ELISA: 1 ?g/ml

Type: Primary
Antigen: Avian Influenza Hemagglutinin
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR