Anti-NCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-188]

Supplier: Abnova
MAB4523
10726-684EA 329 USD
10726-684
Anti-NCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-188]
Antibodies
Mouse monoclonal antibody raised against native NCAM1.

Recommended Dilutions: Flow Cytometry: 10 ?g/ml

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: MEM-188
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Non-human Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR