Anti-CASP6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13138
10705-184EA 428 USD
10705-184
Anti-CASP6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CASP6.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR