Anti-FUT4 Mouse Monoclonal Antibody (Biotin) [clone: MEM-158]

Supplier: Abnova
MAB4531
10715-338EA 329 USD
10715-338
Anti-FUT4 Mouse Monoclonal Antibody (Biotin) [clone: MEM-158]
Antibodies
Mouse monoclonal antibody raised against native FUT4.

Recommended Dilutions: Flow Cytometry: 1:1000

Type: Primary
Antigen: FUT4
Clonality: Biotin
Clone: Monoclonal
Conjugation: MEM-158
Epitope:
Host: IgM
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR