Anti-RHEB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13142
10731-632EA 428 USD
10731-632
Anti-RHEB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of RHEB.

Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: RHEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR