Anti-C1QTNF1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13152
10704-970EA 428 USD
10704-970
Anti-C1QTNF1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of C1QTNF1.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: C1QTNF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR