Anti-C1QTNF5 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13155
10704-150EA 428 USD
10704-150
Anti-C1QTNF5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of C1QTNF5.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: C1QTNF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR