Anti-RB1 Mouse Monoclonal Antibody [clone: 4H6]

Supplier: Abnova
MAB2164
10731-212EA 527.69 USD
10731-212
Anti-RB1 Mouse Monoclonal Antibody [clone: 4H6]
Antibodies
Mouse monoclonal antibody raised against partial recombinant RB1.

Type: Primary
Antigen: RB1
Clonality: Monoclonal
Clone: 4H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR