Anti-MAPK15 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2954
10679-238EA 329 USD
10679-238
Anti-MAPK15 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MAPK15.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-100; Immunohistochemistry: 1:10-50

Type: Primary
Antigen: MAPK15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR