Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-F8 Mouse Monoclonal Antibody

Supplier: Abnova
MAB2175
10627-250
Anti-F8 Mouse Monoclonal Antibody
Mouse monoclonal antibody raised against native F8.

Recommended Dilutions: ELISA: 1:16000

Type: Primary
Antigen: F8
Clonality: Unconjugated
Clone: Monoclonal
Conjugation:
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty