Anti-ABCE1 Goat Polyclonal Antibody

Supplier: Abnova
PAB11421
10687-686EA 406.15 USD
10687-686
Anti-ABCE1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of ABCE1.

Recommended Dilutions: ELISA: 1:32000

Type: Primary
Antigen: ABCE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR