Anti-DPM1 Goat Polyclonal Antibody

Supplier: Abnova
PAB11425
10711-230EA 527.69 USD
10711-230
Anti-DPM1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of Dpm1.

Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: DPM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR