Anti-KLK3 Mouse Monoclonal Antibody [clone: PS2]

Supplier: Abnova
MAB3022
10681-142EA 406.15 USD
10681-142
Anti-KLK3 Mouse Monoclonal Antibody [clone: PS2]
Antibodies
Mouse monoclonal antibody raised against native KLK3.

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: PS2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR