Anti-KLK3 Mouse Monoclonal Antibody [clone: 5A6]

Supplier: Abnova
MAB3025
10682-448EA 406.15 USD
10682-448
Anti-KLK3 Mouse Monoclonal Antibody [clone: 5A6]
Antibodies
Mouse monoclonal antibody raised against native KLK3.

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: 5A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR