Anti-KLK3 Mouse Monoclonal Antibody [clone: 1H12]

Supplier: Abnova
MAB3027
10676-738EA 406.15 USD
10676-738
Anti-KLK3 Mouse Monoclonal Antibody [clone: 1H12]
Antibodies
Mouse monoclonal antibody raised against native KLK3.

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: 1H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR