Anti-OLR1 Mouse Monoclonal Antibody [clone: LOX20-15]

Supplier: Abnova
MAB3041
10676-954EA 786.15 USD
10676-954
Anti-OLR1 Mouse Monoclonal Antibody [clone: LOX20-15]
Antibodies
Mouse monoclonal antibody raised against recombinant OLR1.

Type: Primary
Antigen: OLR1
Clonality: Monoclonal
Clone: LOX20-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR