Anti-TF Mouse Monoclonal Antibody [clone: 8B9]

Supplier: Abnova
MAB3055
10677-338EA 406.15 USD
10677-338
Anti-TF Mouse Monoclonal Antibody [clone: 8B9]
Antibodies
Mouse monoclonal antibody raised against native TF.

Type: Primary
Antigen: TF
Clonality: Monoclonal
Clone: 8B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR