Anti-Troponin I Type 3 Monoclonal Antibody [clone: 22B11]

Supplier: Abnova
MAB3061
10677-384EA 786.15 USD
10677-384
Anti-Troponin I Type 3 Monoclonal Antibody [clone: 22B11]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of TNNI3.

Type: Primary
Antigen: Troponin I Type 3
Clonality: Monoclonal
Clone: 22B11
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR