Anti-ERBB3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12186
10680-942EA 335 USD
10680-942
Anti-ERBB3 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ERBB3.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: ERBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR