Anti-Troponin T type 2 Mouse Monoclonal Antibody [clone: 1F2]

Supplier: Abnova
MAB3089
10677-394EA 527.69 USD
10677-394
Anti-Troponin T type 2 Mouse Monoclonal Antibody [clone: 1F2]
Antibodies
Mouse monoclonal antibody raised against TNNT2.

Type: Primary
Antigen: Troponin T type 2
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR